Learn More
Description
- Hexanoic anhydride has been used in:
- Green synthesis of esters of acyclovir (acyclovir prodrugs)
- Preparation of hexanoyl-modified chitosan nanoparticles and chitosan-based polymeric surfactants via n-acylation of chitosans
Specifications
Specifications
| Molecular Formula | C10H9N3O |
| Linear Formula | C10H9N3O |
| MDL Number | MFCD01443685 |
| Quantity | 250 mL |
| Molecular Weight (g/mol) | 187.2 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.