Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(2-Thienyl)butyric acid, 98%

Catalog No. AAL0756306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0756306 5 g
AAL0756303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0756306 Supplier Thermo Scientific Chemicals Supplier No. L0756306
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4653-11-6 | C8H9O2S | 169.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4653-11-6
MDL Number MFCD00005463

Specifications

Density 1.169
Boiling Point 122°C (0.3 mmHg)
Flash Point >110°C (230°F)
Molecular Formula C11H14O3
Refractive Index 1.532
Quantity 5 g
Beilstein 127982
Synonym 5-4-hydroxyphenyl pentanoic acid, 5-4-hydroxyphenyl valeric acid, pubchem16852, acmc-20am8d, 5-4-hydroxyphenyl pentanoicacid, benzenepentanoic acid, 4-hydroxy
InChI Key YSSJQFONKASLKM-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1CCCCC(=O)O)O
IUPAC Name 5-(4-hydroxyphenyl)pentanoic acid
Molecular Weight (g/mol) 194.23
PubChem CID 7016423
Formula Weight 170.23
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-(2-Thienyl)butyric acid
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement

H315-H319

Causes skin irritation.

Causes serious eye irritation.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 225-090-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.