Learn More
Bio Rad Laboratories 4–20% Criterion TGX Stain-Free Protein Gel, 12+2 well, 45 uL
Catalog No. NC0831741
Encompass
Pkg of 1, 4–20% precast polyacrylamide gel, 13.3 × 8.7 cm (W × L), for use with Criterion and Criterion Dodeca electrophoresis cells and stain-free enabled imagers.
Catalog No. NC0831741 Supplier Bio Rad Laboratories Supplier No. 5678093
Pkg of 1, 4–20% precast polyacrylamide gel, 13.3 × 8.7 cm (W × L), for use with Criterion and Criterion Dodeca electrophoresis cells and stain-free enabled imagers.