Learn More
Bio Rad Laboratories 4-20% CRITERION TGX PRECAST GE
Catalog No. NC0112928
Encompass
These broad range Kaleidoscope prestained standards consist of seven proteins (aprotinin, lysozyme, soybean trypsin inhibitor, carbonic anhydrase, BSA, beta-galactosidase, and myosin) with molecular weights ranging from 7,600 to 216,000.
Catalog No. NC0112928 Supplier Bio Rad Laboratories Supplier No. 5671093
These broad range Kaleidoscope prestained standards consist of seven proteins (aprotinin, lysozyme, soybean trypsin inhibitor, carbonic anhydrase, BSA, beta-galactosidase, and myosin) with molecular weights ranging from 7,600 to 216,000.