Learn More
Bio Rad Laboratories 4-20% CRITERION TGX STAIN-FRE
Catalog No. NC0056287
Encompass
Criterion TGX Stain-Free are long shelf-life Laemmli-like precast polyacrylamide gels for vertical SDS-PAGE and Native PAGE protein separations and for use with Criterion Stain Free Imager. Gel size is 13.3 x 8.7 x 0.1 cm.
Catalog No. NC0056287 Supplier Bio Rad Laboratories Supplier No. 5678095
Criterion TGX Stain-Free are long shelf-life Laemmli-like precast polyacrylamide gels for vertical SDS-PAGE and Native PAGE protein separations and for use with Criterion Stain Free Imager. Gel size is 13.3 x 8.7 x 0.1 cm.