Learn More
Hach Company 4-20 mA Input Board
Supplier:  Hach Company YAB018
Encompass
4-20 mA Input Board for SC1000/SC1500 Controller and 5500 sc AnalyzersAdd additional analog communication capabilities to your controller or analyzer with this 4-20 mA Input card
Catalog No. 50-002-08188