Learn More
Bio Rad Laboratories 4-20% Mini-PROTEAN TGX Stain-Free Protein Gels, 10 well, 30 uL
Catalog No. NC1083979
Encompass
Use these Mini-PROTEAN TGX Stain-Free Protein Gels for separation of polypeptides 2-400 kD.
Catalog No. NC1083979 Supplier Bio Rad Laboratories Supplier No. 4568093
Use these Mini-PROTEAN TGX Stain-Free Protein Gels for separation of polypeptides 2-400 kD.