Learn More
Hach Company 4-20mA Input Module for SC200
Supplier:  Hach Company 9012800
Encompass
4-20mA Input Module for SC200 Universal ControllerThe 4-20mA Input module for the SC200 is a user installed module that allows for input of data from other instruments and electronics via their 4-20mA output to the controller This module works with any 4-20 input from other instruments See manual for additional connection information
Catalog No. 50-002-07419