Learn More
Hach Company 4-20mA Outpt Expan. Mod. SC200
Supplier:  Hach Company 9334600
Encompass
4-20mA Output Expansion Module for SC200 Universal Controller (3 Additional 4-20mA Outputs)When additional communications configurability is needed for process control the 4-20mA output module can be added into the controller to include three additional analog outputs This module connects to the network card on the inside of the SC200
Catalog No. 50-002-07583