Learn More
Chemglass Life Sciences 2.35" ID x 4.28" OD x 1.20" Height White PTFE Support Ring
SDP
Catalog No. 50872569
Encompass_Preferred
2.35" ID x 4.28" OD x 1.20" Height White PTFE Support Ring. PTFE support ring used to support round bottom flasks or cylindrical reaction vessels.PLEASE NOTE: PTFE SUPPORT RINGS CAN BE CUT. PLEASE CALL FOR PRICING
Catalog No. 50-872-569 Supplier Chemglass Life Sciences Supplier No. CG305504
2.35" ID x 4.28" OD x 1.20" Height White PTFE Support Ring. PTFE support ring used to support round bottom flasks or cylindrical reaction vessels.PLEASE NOTE: PTFE SUPPORT RINGS CAN BE CUT. PLEASE CALL FOR PRICING