Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-((2r,3s,4r,5s)-3-(3-chloro-2-fluorophenyl)-4-(4-chloro-2-fluorophenyl)-4-cyano-5-neopentylpyrrolid | 1229705-22-9 | MFCD26142931 | 99.6% | 619.50 | C31H26D3Cl2F2N3O4 | 10 MG
SDP

Catalog No. 5000285850
Encompass_Preferred

Idasanutlin-d3-1 is the deuterium-labeled analogue of idasanutlin supplied as a stable-isotope reference standard for research. It is intended for use as an internal standard or tracer in analytical, pharmacokinetic, and ADME studies of MDM2/p53 inhibitors, enabling accurate quantitation, method validation, and recovery assessment in LC-MS workflows. The product is supplied as a characterized solid with supporting analytical documentation.

  • Used as internal standard for LC-MS quantitation.
  • Suitable for pharmacokinetic and ADME studies.
  • High purity (99.6%).
  • Molecular formula C31H26D3Cl2F2N3O4.
  • Molecular weight 619.50.
  • Supplied with COA, HNMR, LCMS, and SDS.

Catalog No. 50-002-85850 Supplier Medchemexpress LLC Supplier No. HY153939S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Idasanutlin-d3-1 is the deuterium-labeled analogue of idasanutlin supplied as a stable-isotope reference standard for research. It is intended for use as an internal standard or tracer in analytical, pharmacokinetic, and ADME studies of MDM2/p53 inhibitors, enabling accurate quantitation, method validation, and recovery assessment in LC-MS workflows. The product is supplied as a characterized solid with supporting analytical documentation.

  • Used as internal standard for LC-MS quantitation.
  • Suitable for pharmacokinetic and ADME studies.
  • High purity (99.6%).
  • Molecular formula C31H26D3Cl2F2N3O4.
  • Molecular weight 619.50.
  • Supplied with COA, HNMR, LCMS, and SDS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.