Learn More
Medchemexpress LLC 4-(3-(2H-1,2,3-triazol-2-yl)phenylamino)-2-(2-aminocyclohexylamino)pyrimidine-5-carboxamide acetate | 1370261-98-5 | MFCD23105961 | 99.8% | 453.50 | C21H27N9O3 | 10 MG
SDP
Catalog No. 5000285763
Encompass_Preferred
PRT062607 acetate is a research-grade small-molecule inhibitor of spleen tyrosine kinase (Syk), provided as a high-purity solid or as DMSO solutions for laboratory use. It is used in cellular and animal studies to probe Syk-mediated signaling, inflammation, and apoptosis pathways, and is supplied with analytical data to support research applications.
  • Potent and selective Syk inhibition (IC50 ≈ 1 nM).
  • High analytical purity (99.83%).
  • Available as solid powder and 10 mM DMSO solutions for flexibility.
  • Suitable for in vitro and in vivo preclinical studies.
  • Supplied with supporting analytical documentation (COA/SDS).
Catalog No. 50-002-85763 Supplier Medchemexpress LLC Supplier No. HY1532410MG
PRT062607 acetate is a research-grade small-molecule inhibitor of spleen tyrosine kinase (Syk), provided as a high-purity solid or as DMSO solutions for laboratory use. It is used in cellular and animal studies to probe Syk-mediated signaling, inflammation, and apoptosis pathways, and is supplied with analytical data to support research applications.
  • Potent and selective Syk inhibition (IC50 ≈ 1 nM).
  • High analytical purity (99.83%).
  • Available as solid powder and 10 mM DMSO solutions for flexibility.
  • Suitable for in vitro and in vivo preclinical studies.
  • Supplied with supporting analytical documentation (COA/SDS).