Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-[3-[(3-hydroxy-2,4-dioxothieno[3,2-d]pyrimidin-1-yl)methyl]phenyl]benzamide | 824983-94-0 | 99.9% | 100 MG
SDP

Catalog No. 5000230204
Encompass_Preferred

FEN1-IN-2 is a small-molecule inhibitor of flap endonuclease 1 (FEN1) reported to have nanomolar potency and used in biochemical and cellular studies of DNA repair pathways.

  • Nanomolar potency against FEN1 (IC50 3 nM).
  • Activity against XPG (IC50 226 nM).
  • High purity (99.9% by HPLC).
  • Molecular formula C20H15N3O4S and molecular weight 393.42 g/mol.
  • Available in multiple solid pack sizes and as a 10 mM solution in DMSO.

Catalog No. 50-002-30204 Supplier Medchemexpress LLC Supplier No. HY122695100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FEN1-IN-2 is a small-molecule inhibitor of flap endonuclease 1 (FEN1) reported to have nanomolar potency and used in biochemical and cellular studies of DNA repair pathways.

  • Nanomolar potency against FEN1 (IC50 3 nM).
  • Activity against XPG (IC50 226 nM).
  • High purity (99.9% by HPLC).
  • Molecular formula C20H15N3O4S and molecular weight 393.42 g/mol.
  • Available in multiple solid pack sizes and as a 10 mM solution in DMSO.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.