Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(3-{[4-(2-methyl-1H-imidazol-1-yl)phenyl]sulfanyl}phenyl)oxane-4-carboxamide | 179420-17-8 | MFCD00948114 | 98.1% | 393.5 g/mol | C22H23N3O2S | 1 ML
SDP

Catalog No. 5000243165
Encompass_Preferred

CJ-13,610 is a non-redox 5-lipoxygenase (5-LO) inhibitor intended for biochemical and cell-based research. The compound is provided as a ready-to-use 10 mM solution in DMSO or as solid quantities and is formulated for studies of 5-LO pathway modulation.

  • Potent non-redox 5-LO inhibitor suitable for research use.
  • Available as a 10 mM solution in DMSO and as solids in multiple sizes.
  • High purity for reliable experimental results.
  • Molecular formula C22H23N3O2S; molar mass 393.5 g/mol.
  • Recommended storage conditions specified for powder and solution to maintain stability.

Catalog No. 50-002-43165 Supplier Medchemexpress LLC Supplier No. HY10620010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CJ-13,610 is a non-redox 5-lipoxygenase (5-LO) inhibitor intended for biochemical and cell-based research. The compound is provided as a ready-to-use 10 mM solution in DMSO or as solid quantities and is formulated for studies of 5-LO pathway modulation.

  • Potent non-redox 5-LO inhibitor suitable for research use.
  • Available as a 10 mM solution in DMSO and as solids in multiple sizes.
  • High purity for reliable experimental results.
  • Molecular formula C22H23N3O2S; molar mass 393.5 g/mol.
  • Recommended storage conditions specified for powder and solution to maintain stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.