Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-[3-[4-(2-methylimidazol-1-yl)phenyl]sulfanylphenyl]oxane-4-carboxamide | 179420-17-8 | MFCD00948114 | 98.1% | 393.50 g/mol | C22H23N3O2S | 100 MG
SDP

Catalog No. 5000243833
Encompass_Preferred

CJ-13,610 (CAS 179420-17-8) is a small-molecule 5-lipoxygenase inhibitor provided for research use. Supplied as a white to off-white solid and as DMSO stock solutions, it is suitable for in vitro biochemical and cell-based assays probing lipoxygenase pathways.

  • High purity (98.1%).
  • Molecular weight 393.50 g/mol.
  • Chemical formula C22H23N3O2S.
  • White to off-white solid; also available as DMSO solution.
  • Soluble in DMSO at 10 mg/mL; sonication and warming may be required.
  • Storage: powder -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (6 months).

Catalog No. 50-002-43833 Supplier Medchemexpress LLC Supplier No. HY106200100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CJ-13,610 (CAS 179420-17-8) is a small-molecule 5-lipoxygenase inhibitor provided for research use. Supplied as a white to off-white solid and as DMSO stock solutions, it is suitable for in vitro biochemical and cell-based assays probing lipoxygenase pathways.

  • High purity (98.1%).
  • Molecular weight 393.50 g/mol.
  • Chemical formula C22H23N3O2S.
  • White to off-white solid; also available as DMSO solution.
  • Soluble in DMSO at 10 mg/mL; sonication and warming may be required.
  • Storage: powder -20 °C (3 years) or 4 °C (2 years); in solvent -80 °C (6 months).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.