Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-[3-[4-(2-methylimidazol-1-yl)phenyl]sulfanylphenyl]oxane-4-carboxamide | 179420-17-8 | MFCD00948114 | 98.1% | 393.50 g/mol | C22H23N3O2S | 25 MG
SDP

Catalog No. 5000242941
Encompass_Preferred

CJ-13,610 is a small-molecule 5-lipoxygenase (5-LO) inhibitor supplied for research use. It is isolated as a white to off-white solid with high analytical purity and is intended for biochemical and pharmacological studies of lipoxygenase pathways.

  • High purity (98.1%) suitable for research applications.
  • Molecular formula C22H23N3O2S and molecular weight 393.50 g/mol.
  • Soluble in DMSO at approximately 10 mg/mL with warming or sonication.
  • Powder stability: -20°C for up to 3 years; refrigerated stability 2 years at 4°C.
  • Available in multiple premeasured quantities and as DMSO solutions for convenience.

Catalog No. 50-002-42941 Supplier Medchemexpress LLC Supplier No. HY10620025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CJ-13,610 is a small-molecule 5-lipoxygenase (5-LO) inhibitor supplied for research use. It is isolated as a white to off-white solid with high analytical purity and is intended for biochemical and pharmacological studies of lipoxygenase pathways.

  • High purity (98.1%) suitable for research applications.
  • Molecular formula C22H23N3O2S and molecular weight 393.50 g/mol.
  • Soluble in DMSO at approximately 10 mg/mL with warming or sonication.
  • Powder stability: -20°C for up to 3 years; refrigerated stability 2 years at 4°C.
  • Available in multiple premeasured quantities and as DMSO solutions for convenience.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.