Learn More
Cayman Chemical 4-(3,4-Difluorobenzo)curcumin, Purity: ≥95%, CAS: 1170354-22-9, 25 mg
SDP
Catalog No. 502918409
Encompass_Preferred
A semisynthetic coumarin with antiparasitic and anticancer activities; reduces the number of Vero cells infected with T. gondii promastigotes and amastigotes (EC50s = 0.8 and 0.37 µM, respectively); enhances 5-fluorouracil- and oxaliplatin-induced inhibition of primary and secondary colonosphere formation by chemoresistant HCT116 cells at 4 and 8 µM
Catalog No. 50-291-8409 Supplier Cayman Chemical Supplier No. 3927125 MG
A semisynthetic coumarin with antiparasitic and anticancer activities; reduces the number of Vero cells infected with T. gondii promastigotes and amastigotes (EC50s = 0.8 and 0.37 µM, respectively); enhances 5-fluorouracil- and oxaliplatin-induced inhibition of primary and secondary colonosphere formation by chemoresistant HCT116 cells at 4 and 8 µM