Learn More
Medchemexpress LLC 4-(3,4-dihydroisoquinolin-2(1H)-yl)-2-(pyridin-3-yl)quinazoline | 796079-91-9 | 97.0% | 338.41 g/mol | C22H18N4 | 10 MG
SDP
Catalog No. 5000288038
Encompass_Preferred
GCase modulator-1 (compound 9g) is a quinazoline-derivative small molecule reported to modulate glucocerebrosidase (GCase) activity (reported AC50 = 2.23 μM). It is supplied as a research-grade powder for use in biochemical assays and in vitro studies; reported molecular weight is 338.41 g/mol and reported purity is ~97%.
  • Modulates GCase (glucosidase) with reported AC50 = 2.23 μM.
  • Quinazoline-derived small-molecule scaffold suitable for biochemical studies.
  • Provided as a powder for preparation of assay solutions.
  • Reported purity approximately 97% and molecular weight 338.41 g/mol.
  • Intended for research use in vitro; not for human or clinical use.
Catalog No. 50-002-88038 Supplier Medchemexpress LLC Supplier No. HY15628710MG
GCase modulator-1 (compound 9g) is a quinazoline-derivative small molecule reported to modulate glucocerebrosidase (GCase) activity (reported AC50 = 2.23 μM). It is supplied as a research-grade powder for use in biochemical assays and in vitro studies; reported molecular weight is 338.41 g/mol and reported purity is ~97%.
  • Modulates GCase (glucosidase) with reported AC50 = 2.23 μM.
  • Quinazoline-derived small-molecule scaffold suitable for biochemical studies.
  • Provided as a powder for preparation of assay solutions.
  • Reported purity approximately 97% and molecular weight 338.41 g/mol.
  • Intended for research use in vitro; not for human or clinical use.