Learn More
Medchemexpress LLC 4-[3-(4-hydroxybutyl)-4,4-dimethyl-2,5-dioxoimidazolidin-1-yl]-2-(trifluoromethyl)benzonitrile | 154992-24-2 | MFCD00925776 | 99.7% | 369.34 g/mol | C17H18F3N3O3 | 5 MG
SDP
Catalog No. 5000240245
Encompass_Preferred
RU 58841 is a nonsteroidal androgen receptor antagonist provided for research use. It acts as a specific anti-androgen with reported effects on hair regrowth, and is supplied as a high-purity powder with documented storage and stability recommendations.
  • Specific androgen receptor antagonist for in vitro and in vivo research.
  • High declared purity greater than 99.7%.
  • Available in small research quantities such as 5 mg powder.
  • Powder form with recommended storage to maintain stability.
  • Molecular weight 369.34 g/mol; molecular formula C17H18F3N3O3.
Catalog No. 50-002-40245 Supplier Medchemexpress LLC Supplier No. HY105615MG
RU 58841 is a nonsteroidal androgen receptor antagonist provided for research use. It acts as a specific anti-androgen with reported effects on hair regrowth, and is supplied as a high-purity powder with documented storage and stability recommendations.
  • Specific androgen receptor antagonist for in vitro and in vivo research.
  • High declared purity greater than 99.7%.
  • Available in small research quantities such as 5 mg powder.
  • Powder form with recommended storage to maintain stability.
  • Molecular weight 369.34 g/mol; molecular formula C17H18F3N3O3.