Learn More
Sklar Instruments Single Use, Econo-Sterile, Adson Dressing Forceps, Sklar 4-3/4 in., Blunt, Straight, Serrated, Stainless Steel, Disposable, CS of 50. Adson Dressing Forceps are thumb forceps.
SDP
Catalog No. 501094282 Shop All Sklar Instruments Products
Encompass_Preferred
Single Use, Econo-Sterile, Adson Dressing Forceps, Sklar, 4-3/4 in., Blunt, Straight, Serrated, Stainless Steel, Disposable, CS of 50. Adson Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds.
Catalog No. 50-109-4282 Supplier Sklar Instruments Supplier No. 962570
Single Use, Econo-Sterile, Adson Dressing Forceps, Sklar, 4-3/4 in., Blunt, Straight, Serrated, Stainless Steel, Disposable, CS of 50. Adson Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds.