Learn More
Sklar Instruments Single Use, Econo-Sterile, Derf Needle Holder, Sklar 4-3/4 in., Straight, Serrated with Groove, Stainless Steel, Disposable. Box of 50.
SDP
Catalog No. 501094292 Shop All Sklar Instruments Products
Encompass_Preferred
Single Use, Econo-Sterile, Derf Needle Holder, Sklar, 4-3/4 in., Straight, Serrated with Groove, Stainless Steel, Disposable. Box of 50. Derf Needle Holders feature short jaws and delicate serrations that hold small needles and commonly 5-0 and 6-0 sutures.
Catalog No. 50-109-4292 Supplier Sklar Instruments Supplier No. 962582
Single Use, Econo-Sterile, Derf Needle Holder, Sklar, 4-3/4 in., Straight, Serrated with Groove, Stainless Steel, Disposable. Box of 50. Derf Needle Holders feature short jaws and delicate serrations that hold small needles and commonly 5-0 and 6-0 sutures.