Learn More
Sklar Instruments Single Use, Econo-Sterile, Wire Cutting Scissors, Sklar 4-3/4 in., Blunt/Blunt, Angled, Serrated, Stainless Steel, CS of 25.
SDP
Catalog No. 501094237 Shop All Sklar Instruments Products
Encompass_Preferred
Single Use, Econo-Sterile, Wire Cutting Scissors, Sklar, 4-3/4 in., Blunt/Blunt, Angled, Serrated, Stainless Steel, CS of 25.
Catalog No. 50-109-4237 Supplier Sklar Instruments Supplier No. 962519
Single Use, Econo-Sterile, Wire Cutting Scissors, Sklar, 4-3/4 in., Blunt/Blunt, Angled, Serrated, Stainless Steel, CS of 25.