Learn More
Medchemexpress LLC 4-(3-(5-chloro-2-methoxyphenyl)thioureido)-1-ethyl-1H-pyrazole-3-carboxamide | 957485-64-2 | 99.3% | 25 MG
SDP
Catalog No. 5000231982
Encompass_Preferred
FPH2 is a small-molecule research compound that induces functional proliferation of primary human hepatocytes and is used for studies of liver biology and early therapeutic development.
  • Induces functional proliferation of primary human hepatocytes.
  • Used for research into liver regeneration and disease models.
  • High purity suitable for research applications.
  • White to off-white solid for convenient handling.
  • Stable when stored frozen as a powder or in solvent under recommended conditions.
  • Molecular formula C14H16ClN5O2S and molecular weight 353.83 g/mol.
Catalog No. 50-002-31982 Supplier Medchemexpress LLC Supplier No. HY1228125MG
FPH2 is a small-molecule research compound that induces functional proliferation of primary human hepatocytes and is used for studies of liver biology and early therapeutic development.
  • Induces functional proliferation of primary human hepatocytes.
  • Used for research into liver regeneration and disease models.
  • High purity suitable for research applications.
  • White to off-white solid for convenient handling.
  • Stable when stored frozen as a powder or in solvent under recommended conditions.
  • Molecular formula C14H16ClN5O2S and molecular weight 353.83 g/mol.