Learn More
Medchemexpress LLC 4-[(3,5-diamino-1H-pyrazol-4-yl)azo]-phenol | 140651-18-9 | MFCD09262258 | 99.6% | 218.22 g/mol | C9H10N6O | 50 MG
SDP
Catalog No. 5000214046
Encompass_Preferred
CAN508 is a potent, ATP-competitive inhibitor of cyclin-dependent kinase 9 (CDK9)/cyclin T1 intended for research use. It inhibits CDK9/cyclin T1 with an IC50 of 0.35 μM, displays ~38-fold selectivity over other CDK/cyclin complexes, and has demonstrated antitumor activity in preclinical studies.
  • Potent ATP-competitive inhibitor of CDK9/cyclin T1 (IC50 0.35 μM).
  • Approximately 38-fold selectivity versus other CDK/cyclin complexes.
  • Reported antitumor activity in preclinical models.
  • High purity, 99.6%.
  • Molecular weight 218.22 g/mol; formula C9H10N6O.
  • Supplied with supporting documentation such as SDS, COA, and LCMS.
  • Intended for research use only.
Catalog No. 50-002-14046 Supplier Medchemexpress LLC Supplier No. HY10042950MG
CAN508 is a potent, ATP-competitive inhibitor of cyclin-dependent kinase 9 (CDK9)/cyclin T1 intended for research use. It inhibits CDK9/cyclin T1 with an IC50 of 0.35 μM, displays ~38-fold selectivity over other CDK/cyclin complexes, and has demonstrated antitumor activity in preclinical studies.
  • Potent ATP-competitive inhibitor of CDK9/cyclin T1 (IC50 0.35 μM).
  • Approximately 38-fold selectivity versus other CDK/cyclin complexes.
  • Reported antitumor activity in preclinical models.
  • High purity, 99.6%.
  • Molecular weight 218.22 g/mol; formula C9H10N6O.
  • Supplied with supporting documentation such as SDS, COA, and LCMS.
  • Intended for research use only.