Learn More
Medchemexpress LLC 4-[(3,5-dichloro-4-methoxyphenyl)amino]-6-methoxy-7-[3-(4-Me-piperazinyl)propoxy]quinoline-3-carbonitrile | 1391063-17-4 | MFCD22418831 | 99.6% | 530.45 g/mol | C26H29Cl2N5O3 | 5 MG
SDP
Catalog No. 5000245953
Encompass_Preferred
Bosutinib isomer (CAS 1391063-17-4) is a small-molecule inhibitor of the cell-cycle kinases Wee1 and Wee2, with reported Kd values of 43.7 ± 10.0 nM (Wee1) and 4.7 ± 2.3 nM (Wee2). The compound has molecular formula C26H29Cl2N5O3, molecular weight 530.45 g/mol, and is characterized by high LCMS purity.
  • High affinity for Wee1 and Wee2 kinases.
  • Reported Kd: 43.7 ± 10.0 nM (Wee1); 4.7 ± 2.3 nM (Wee2).
  • Molecular formula C26H29Cl2N5O3; MW 530.45 g/mol.
  • High purity by LCMS (99.62%).
  • Available in small research quantities, including 5 MG vials.
  • Datasheet and COA provided for characterization data.
Catalog No. 50-002-45953 Supplier Medchemexpress LLC Supplier No. HY1030295MG
Bosutinib isomer (CAS 1391063-17-4) is a small-molecule inhibitor of the cell-cycle kinases Wee1 and Wee2, with reported Kd values of 43.7 ± 10.0 nM (Wee1) and 4.7 ± 2.3 nM (Wee2). The compound has molecular formula C26H29Cl2N5O3, molecular weight 530.45 g/mol, and is characterized by high LCMS purity.
  • High affinity for Wee1 and Wee2 kinases.
  • Reported Kd: 43.7 ± 10.0 nM (Wee1); 4.7 ± 2.3 nM (Wee2).
  • Molecular formula C26H29Cl2N5O3; MW 530.45 g/mol.
  • High purity by LCMS (99.62%).
  • Available in small research quantities, including 5 MG vials.
  • Datasheet and COA provided for characterization data.