Learn More
Medchemexpress LLC 4-(3,5-difluoro-4-hydroxybenzylidene)-1-methyl-5-oxo-4,5-dihydro-1H-imidazole-2-carbaldehyde oxime | 1420815-34-4 | 99.8% | 281.21 g·mol-1 | C12H9F2N3O3 | 5 MG
SDP
Catalog No. 5000222105
Encompass_Preferred
DFHO is a fluorogenic small molecule that is activated by binding to the Corn RNA aptamer, converting into a fluorescent form and enabling RNA imaging in living cells. It exhibits low background fluorescence and low cytotoxicity, and is supplied at high purity for research applications.
  • Binds the Corn RNA aptamer and becomes fluorescent (Kd ≈ 70 nM).
  • Enables live-cell RNA imaging via aptamer-activated fluorescence.
  • Low background fluorescence and low cytotoxicity in cellular assays.
  • High purity suitable for biochemical and imaging experiments.
  • Supplied in small-mass pack sizes for research use.
Catalog No. 50-002-22105 Supplier Medchemexpress LLC Supplier No. HY1362775MG
DFHO is a fluorogenic small molecule that is activated by binding to the Corn RNA aptamer, converting into a fluorescent form and enabling RNA imaging in living cells. It exhibits low background fluorescence and low cytotoxicity, and is supplied at high purity for research applications.
  • Binds the Corn RNA aptamer and becomes fluorescent (Kd ≈ 70 nM).
  • Enables live-cell RNA imaging via aptamer-activated fluorescence.
  • Low background fluorescence and low cytotoxicity in cellular assays.
  • High purity suitable for biochemical and imaging experiments.
  • Supplied in small-mass pack sizes for research use.