Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(3,6-Dimethyl-3-heptyl)phenol-ring-13C6 Solution, 100 μg/mL in Acetone, MilliporeSigma™ Supelco™

Catalog No. 111017949 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7949 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7949 Supplier MilliporeSigma Supelco Supplier No. 324711ML
Add to Cart
Add to Cart

Certan Vial

Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 1173020-38-6
Flash Point -17.0°C - Closed Cup
Linear Formula 13C6C9H24 O
MDL Number MFCD09842832
Synonym 3,6,3-Nonylphenol-13C6; 363-NP-13C6; 4-(1-Ethyl-1,4-dimethylpentyl)phenol-13C6 (ring-13C6)
Type Analytical Standard
Quantity 1 mL
Formula Weight 226.31
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 100 μg/mL in acetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.