Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-[3-acetyl-2,5-dihydro-4-hydroxy-1-[2-(2-methyl-1H-indol-3-yl)ethyl]-5-oxo-1H-pyrrol-2-yl]-benzoic | 1560894-05-4 | 98.1% | 432.47 | C25H24N2O5 | 5 MG
SDP

Catalog No. 5000226467
Encompass_Preferred

PYD-106 is a stereoselective pyrrolidinone positive allosteric modulator selective for GluN2C-containing NMDA receptors, provided as a high-purity research chemical. It increases channel opening frequency and is intended for laboratory research use only.

  • Selective positive allosteric modulator of GluN2C-containing NMDA receptors.
  • Stereoselective pyrrolidinone scaffold with defined pharmacological activity.
  • High purity (98.13%) suitable for research applications.
  • Available in small research sizes, including 5 mg.
  • Recommended storage: powder at -20°C for long-term stability.
  • Chemical formula C25H24N2O5; molecular weight 432.47.

Catalog No. 50-002-26467 Supplier Medchemexpress LLC Supplier No. HY1177345MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PYD-106 is a stereoselective pyrrolidinone positive allosteric modulator selective for GluN2C-containing NMDA receptors, provided as a high-purity research chemical. It increases channel opening frequency and is intended for laboratory research use only.

  • Selective positive allosteric modulator of GluN2C-containing NMDA receptors.
  • Stereoselective pyrrolidinone scaffold with defined pharmacological activity.
  • High purity (98.13%) suitable for research applications.
  • Available in small research sizes, including 5 mg.
  • Recommended storage: powder at -20°C for long-term stability.
  • Chemical formula C25H24N2O5; molecular weight 432.47.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.