Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(3-Aminopropyl)thiomorpholine 1,1-Dioxide 98.0+%, TCI America™
SDP

Catalog No. A18041G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18041G 1 g
A18045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A18041G Supplier TCI America Supplier No. A18041G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 90000-25-2
Molecular Formula C7H16N2O2S
Molecular Weight (g/mol) 192.28
MDL Number MFCD03840683
InChI Key FSRMWCVZAUEPEQ-UHFFFAOYSA-N
Synonym 3-(1,1-Dioxothiomorpholino)propylamine
PubChem CID 24728054
IUPAC Name 4-(3-aminopropyl)-1λ⁶-thiomorpholine-1,1-dione
SMILES NCCCN1CCS(=O)(=O)CC1

Specifications

Color White-Yellow
Boiling Point 190°C
Quantity 1 g
Formula Weight 192.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-(3-Aminopropyl)thiomorpholine 1,1-Dioxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.