Learn More
Medchemexpress LLC 4-(3-butoxy-4-methoxybenzyl)-2-imidazolidinone | 29925-17-5 | 100.0% | 278.35 g/mol | C15H22N2O3 | 50 MG
SDP
Catalog No. 5000212621
Encompass_Preferred
Ro 20-1724 is a small-molecule research reagent that inhibits cAMP-specific phosphodiesterase 4 (PDE4). It is commonly used in preclinical studies to probe PDE4-mediated signaling and neuroprotective mechanisms; the compound is reported with a Ki of 1,930 nM.
Catalog No. 50-002-12621 Supplier Medchemexpress LLC Supplier No. HY10092750MG
Ro 20-1724 is a small-molecule research reagent that inhibits cAMP-specific phosphodiesterase 4 (PDE4). It is commonly used in preclinical studies to probe PDE4-mediated signaling and neuroprotective mechanisms; the compound is reported with a Ki of 1,930 nM.