Learn More
Medchemexpress LLC 4-(3-butoxy-4-methoxybenzyl)imidazolidin-2-one | 29925-17-5 | MFCD00077397 | 100.0% | 278.35 g·mol⁻¹ | C15H22N2O3 | 1 ML
SDP
Catalog No. 5000212626
Encompass_Preferred
Ro 20-1724 is a small-molecule inhibitor of cAMP-specific phosphodiesterase 4 (PDE4) used in in vitro research. It has a reported Ki of 1,930 nM and is supplied as a 10 mM solution in DMSO for convenient dosing in biochemical and cell-based assays.
  • Potent PDE4 inhibitor with reported Ki of 1,930 nM.
  • Provided as a 10 mM solution in DMSO (1 mL) for easy handling.
  • High purity suitable for research applications (reported 99.98%).
  • Molecular weight 278.35 g·mol⁻¹ and formula C15H22N2O3 for accurate stoichiometry.
  • Documented neuroprotective activity in literature, useful for mechanism studies.
  • Stable in solvent under recommended storage conditions.
Catalog No. 50-002-12626 Supplier Medchemexpress LLC Supplier No. HY10092710MM 1ML
Ro 20-1724 is a small-molecule inhibitor of cAMP-specific phosphodiesterase 4 (PDE4) used in in vitro research. It has a reported Ki of 1,930 nM and is supplied as a 10 mM solution in DMSO for convenient dosing in biochemical and cell-based assays.
  • Potent PDE4 inhibitor with reported Ki of 1,930 nM.
  • Provided as a 10 mM solution in DMSO (1 mL) for easy handling.
  • High purity suitable for research applications (reported 99.98%).
  • Molecular weight 278.35 g·mol⁻¹ and formula C15H22N2O3 for accurate stoichiometry.
  • Documented neuroprotective activity in literature, useful for mechanism studies.
  • Stable in solvent under recommended storage conditions.