Learn More
Medchemexpress LLC 4-(3-butoxy-4-methoxybenzyl)-imidazolidin-2-one | 29925-17-5 | MFCD00077397 | 100.0% | 278.35 g/mol | C15H22N2O3 | 100 MG
SDP
Catalog No. 5000213109
Encompass_Preferred
Ro 20-1724 is a research-grade small molecule inhibitor of cAMP-specific phosphodiesterase 4 (PDE4). It has a reported Ki of 1,930 nM and has been used in studies demonstrating neuroprotective effects. The compound is supplied with high chemical purity and is intended for biochemical assays and preclinical research.
  • Inhibits cAMP-specific phosphodiesterase 4 (PDE4).
  • Reported Ki of 1,930 nM against PDE4.
  • High purity suitable for research applications.
  • Available as solid and as DMSO solution formats.
  • Commonly used in enzymology and neuroprotection studies.
Catalog No. 50-002-13109 Supplier Medchemexpress LLC Supplier No. HY100927100MG
Ro 20-1724 is a research-grade small molecule inhibitor of cAMP-specific phosphodiesterase 4 (PDE4). It has a reported Ki of 1,930 nM and has been used in studies demonstrating neuroprotective effects. The compound is supplied with high chemical purity and is intended for biochemical assays and preclinical research.
  • Inhibits cAMP-specific phosphodiesterase 4 (PDE4).
  • Reported Ki of 1,930 nM against PDE4.
  • High purity suitable for research applications.
  • Available as solid and as DMSO solution formats.
  • Commonly used in enzymology and neuroprotection studies.