Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(3-carboxyphenyl)-2,5-pyridinedicarboxylic acid | 29553-70-6 | 99.6% | 287.22 | C14H9NO6 | 25MG
SDP

Catalog No. 5000235447
Encompass_Preferred

4-CPPC is a small-molecule research reagent that functions as a reversible, selective inhibitor of macrophage migration inhibitory factor-2 (MIF-2). It is intended for in vitro biochemical and cell-based research and is supplied with characterization and high reported purity; not for human use.

  • Reversible, selective MIF-2 inhibitor with reported Ki/IC50 values of 431 μM for MIF-1 and 33 μM for MIF-2.
  • High purity (99.6%) suitable for research applications.
  • Molecular formula C14H9NO6 and molecular weight 287.22 g/mol.
  • Soluble in DMSO; also offered as a 10 mM DMSO solution option.
  • Available in multiple small pack sizes for laboratory use.
  • For research use only, not intended for human or clinical use.

Catalog No. 50-002-35447 Supplier Medchemexpress LLC Supplier No. HY12381425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-CPPC is a small-molecule research reagent that functions as a reversible, selective inhibitor of macrophage migration inhibitory factor-2 (MIF-2). It is intended for in vitro biochemical and cell-based research and is supplied with characterization and high reported purity; not for human use.

  • Reversible, selective MIF-2 inhibitor with reported Ki/IC50 values of 431 μM for MIF-1 and 33 μM for MIF-2.
  • High purity (99.6%) suitable for research applications.
  • Molecular formula C14H9NO6 and molecular weight 287.22 g/mol.
  • Soluble in DMSO; also offered as a 10 mM DMSO solution option.
  • Available in multiple small pack sizes for laboratory use.
  • For research use only, not intended for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.