Learn More
Medchemexpress LLC 4-(3-carboxyphenyl)-2,5-pyridinedicarboxylic acid | 29553-70-6 | 99.6% | 287.22 | C14H9NO6 | 5MG
SDP
Catalog No. 5000236831
Encompass_Preferred
4-CPPC is a small-molecule research reagent that acts as a potent, reversible, and selective inhibitor of macrophage migration inhibitory factor 2 (MIF-2). It shows Ki values of 431 μM for MIF-1 and 33 μM for MIF-2 and is supplied as a solid with reported high purity.
Catalog No. 50-002-36831 Supplier Medchemexpress LLC Supplier No. HY1238145MG
4-CPPC is a small-molecule research reagent that acts as a potent, reversible, and selective inhibitor of macrophage migration inhibitory factor 2 (MIF-2). It shows Ki values of 431 μM for MIF-1 and 33 μM for MIF-2 and is supplied as a solid with reported high purity.