Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(3-Chloro-4-(3-(cyclopropyl-2,2,3,3-d4)ureido)phenoxy)-7-methoxyquinoline-6-carboxamide-15N | 98.0% | 431.87 | 10 MG
SDP

Catalog No. 5000344353
Encompass_Preferred

Lenvatinib-15N,d4 is a chemical compound with a molecular weight of 431.87 and a purity of 98.01% as determined by LCMS. It is presented as a white to off-white solid. This product has not been fully validated for medical applications and is intended for research use only.

  • Appearance: White to off-white solid
  • Storage as powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
  • ¹H NMR spectrum is consistent with structure
  • LCMS is consistent with structure
  • Complies with given specifications

Catalog No. 50-003-44353 Supplier Medchemexpress LLC Supplier No. HY10981S210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lenvatinib-15N,d4 is a chemical compound with a molecular weight of 431.87 and a purity of 98.01% as determined by LCMS. It is presented as a white to off-white solid. This product has not been fully validated for medical applications and is intended for research use only.

  • Appearance: White to off-white solid
  • Storage as powder: -20°C for 3 years, 4°C for 2 years
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month
  • ¹H NMR spectrum is consistent with structure
  • LCMS is consistent with structure
  • Complies with given specifications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.