Learn More
Medchemexpress LLC 4-(3-Chloro-4-(3-(cyclopropyl-2,2,3,3-d4)ureido)phenoxy)-7-methoxyquinoline-6-carboxamide-15N | 98.01% | 431.87 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY10981S2
Encompass_Preferred
Lenvatinib-15N,d4 is a 15N and deuterated labeled version of Lenvatinib. Lenvatinib (E7080) is an oral, multi-targeted tyrosine kinase inhibitor that inhibits VEGFR1-3, FGFR1-4, PDGFR, KIT, and RET, and exhibits potent antitumor activities.
  • Isotope-labeled compound
  • Deuterated and 15N labeled
  • Oral, multi-targeted tyrosine kinase inhibitor
  • Inhibits VEGFR1-3, FGFR1-4, PDGFR, KIT, and RET
  • Shows potent antitumor activities
Catalog No. 50-003-47675