Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(3-chloro-4-cyanophenyl)-2-(2-((1R,2R)-2-hydroxy-2-methylcyclopropyl)amino)pyrimidi. | 1610677-37-6 | 99.6% | 535.04 g·mol⁻1 | C28H31ClN6O3 | 1 ML
SDP

Catalog No. 5000211960
Encompass_Preferred

Luvixasertib hydrochloride is a potent, selective inhibitor of the TTK/Mps1 kinase with low-nanomolar biochemical potency and demonstrated in vitro anticancer activity. Supplied as the hydrochloride salt in a ready-to-use 10 mM DMSO solution, it is intended for preclinical kinase assays and cellular studies probing mitotic checkpoint biology.

  • High potency: IC50 1.7 nM for TTK in vitro.
  • Hydrochloride salt suitable for formulation and storage.
  • Provided as a 10 mM solution in DMSO, 1 mL ready to use.
  • High purity (99.63% by supplier analysis).
  • Well characterized: CAS 1610677-37-6; molecular weight 535.04 g·mol⁻1.
  • Applications: preclinical kinase inhibition and cellular mitotic checkpoint studies.

Catalog No. 50-002-11960 Supplier Medchemexpress LLC Supplier No. HY101340A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Luvixasertib hydrochloride is a potent, selective inhibitor of the TTK/Mps1 kinase with low-nanomolar biochemical potency and demonstrated in vitro anticancer activity. Supplied as the hydrochloride salt in a ready-to-use 10 mM DMSO solution, it is intended for preclinical kinase assays and cellular studies probing mitotic checkpoint biology.

  • High potency: IC50 1.7 nM for TTK in vitro.
  • Hydrochloride salt suitable for formulation and storage.
  • Provided as a 10 mM solution in DMSO, 1 mL ready to use.
  • High purity (99.63% by supplier analysis).
  • Well characterized: CAS 1610677-37-6; molecular weight 535.04 g·mol⁻1.
  • Applications: preclinical kinase inhibition and cellular mitotic checkpoint studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.