Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(3-chloro-4-fluoroanilino)-6-(pyridin-3-ylmethylamino)-1,7-naphthyridine-3-carbonitrile | 871307-18-5 | MFCD10566771 | 99.4% | 404.83 g/mol | C21H14ClFN6 | 50MG
SDP

Catalog No. 5000236702
Encompass_Preferred

Tpl2 Kinase Inhibitor 1 is a small-molecule, ATP-competitive inhibitor of the Tpl2 (MAP3K8/COT) kinase with demonstrated biochemical and cellular activity. It has a biochemical IC50 of 50 nM and reduces LPS-induced TNF-α production in cellular assays. The compound is supplied as a purified solid and is also available as a 10 mM solution in DMSO for research applications.

  • Selective Tpl2 (MAP3K8) inhibitor with biochemical IC50 = 50 nM.
  • Reduces LPS-induced TNF-α production in cellular assays (cellular IC50s reported).
  • Molecular formula C21H14ClFN6; molecular weight 404.83 g/mol.
  • Purity 99.42% as listed by supplier.
  • Available as solid in multiple quantities and as 10 mM in 1 mL DMSO.
  • Suitable for in vitro and cellular research applications.

Catalog No. 50-002-36702 Supplier Medchemexpress LLC Supplier No. HY1235850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tpl2 Kinase Inhibitor 1 is a small-molecule, ATP-competitive inhibitor of the Tpl2 (MAP3K8/COT) kinase with demonstrated biochemical and cellular activity. It has a biochemical IC50 of 50 nM and reduces LPS-induced TNF-α production in cellular assays. The compound is supplied as a purified solid and is also available as a 10 mM solution in DMSO for research applications.

  • Selective Tpl2 (MAP3K8) inhibitor with biochemical IC50 = 50 nM.
  • Reduces LPS-induced TNF-α production in cellular assays (cellular IC50s reported).
  • Molecular formula C21H14ClFN6; molecular weight 404.83 g/mol.
  • Purity 99.42% as listed by supplier.
  • Available as solid in multiple quantities and as 10 mM in 1 mL DMSO.
  • Suitable for in vitro and cellular research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.