Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-[(3-chloro-4-fluorophenyl)amino]-6-[(3-pyridinylmethyl)amino]-1,7-naphthyridine-3-carbonitrile | 871307-18-5 | 99.4% | 404.83 g/mol | C21H14ClFN6 | 100MG
SDP

Catalog No. 5000235975
Encompass_Preferred

Tpl2 Kinase Inhibitor 1 is a selective small-molecule inhibitor of Tpl2 (MAP3K8) used for biochemical and cell-based research. It demonstrates potent biochemical inhibition and documented cellular effects, and is supplied as a solid with defined purity and storage conditions for laboratory use.

  • Biochemical IC50 ≈ 50 nM for Tpl2 (MAP3K8).
  • Molecular weight 404.83 g/mol.
  • Chemical formula C21H14ClFN6.
  • Purity 99.4% (reported by manufacturer).
  • Appearance solid, yellow to green.
  • Available in solid sizes up to 100 mg.
  • Recommended storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Documented cellular activity: <5 μM (A-431 cells) and 1.1 μM (monocytes).

Catalog No. 50-002-35975 Supplier Medchemexpress LLC Supplier No. HY12358100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Tpl2 Kinase Inhibitor 1 is a selective small-molecule inhibitor of Tpl2 (MAP3K8) used for biochemical and cell-based research. It demonstrates potent biochemical inhibition and documented cellular effects, and is supplied as a solid with defined purity and storage conditions for laboratory use.

  • Biochemical IC50 ≈ 50 nM for Tpl2 (MAP3K8).
  • Molecular weight 404.83 g/mol.
  • Chemical formula C21H14ClFN6.
  • Purity 99.4% (reported by manufacturer).
  • Appearance solid, yellow to green.
  • Available in solid sizes up to 100 mg.
  • Recommended storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Documented cellular activity: <5 μM (A-431 cells) and 1.1 μM (monocytes).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.