Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

{4-[3-(Dimethylamino)propoxy]phenyl}methanol, 97%, Thermo Scientific™

Catalog No. CC59509DA
Change view
Click to view available options
:
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
CC59509DA Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. CC59509DA Supplier Thermo Scientific Chemicals Supplier No. CC59509DA
Only null left
Add to Cart
This item is not returnable. View return policy

Specifications

CAS 426831-08-5
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C12H19NO2
Quantity 1 g
MDL Number MFCD06202771
Synonym 4-3-dimethylamino propoxy phenyl methanol, benzenemethanol,4-3-dimethylamino propoxy, 4-3-dimethylamino propoxy phenyl methan-1-ol
InChI Key ZLCIFBOMXUAJPZ-UHFFFAOYSA-N
SMILES CN(C)CCCOC1=CC=C(C=C1)CO
IUPAC Name [4-[3-(dimethylamino)propoxy]phenyl]methanol
Molecular Weight (g/mol) 209.289
PubChem CID 9920748
Formula Weight 209.29
Percent Purity 97%
Chemical Name or Material {4-[3-(Dimethylamino)propoxy]phenyl}methanol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.