Learn More
Medchemexpress LLC 4-(3-Fluorobenzyloxy)benzaldehyde | 66742-57-2 | 99.8% | C14H11FO2 | 100 G
SDP
Catalog No. 5000434174
Encompass_Preferred
4-(3-Fluorobenzyloxy)benzaldehyde is a chemical compound that serves as a drug intermediate. It is used in the synthesis of various active compounds for research purposes.
  • Solid appearance
  • Functions as a drug intermediate
  • Suitable for the synthesis of active compounds
  • For research use only
  • Shipped at room temperature in continental US
  • Store at 4°C under nitrogen
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen
Catalog No. 50-004-34174 Supplier Medchemexpress LLC Supplier No. HY76181100G
4-(3-Fluorobenzyloxy)benzaldehyde is a chemical compound that serves as a drug intermediate. It is used in the synthesis of various active compounds for research purposes.
  • Solid appearance
  • Functions as a drug intermediate
  • Suitable for the synthesis of active compounds
  • For research use only
  • Shipped at room temperature in continental US
  • Store at 4°C under nitrogen
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month under nitrogen