Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-(3-Methoxy-benzyl)-piperidine

Catalog No. BBO0004371G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BBO0004371G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BBO0004371G Supplier Sigma Aldrich Supplier No. BBO0004371G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Reactant involved in:
  • Cyclocondensation and deacetylation of dibromo(phenylsulfonyl)propene derivatives for synthesis of disubstituted furans
  • Synthesis of dual activity effectors of angiotensin ii type 1 receptors and peroxisome proliferator-activated receptor-α
  • Addition reactions with 1-alkynes
  • Knoevenagel condensation
  • Oxidative free radical reactions with 2-amino-1,4-benzoquinones
  • Mild oxidative cleavage for synthesis of carboxylic acids

Specifications

Molecular Formula C13H19NO
Linear Formula C13H19NO
MDL Number MFCD05863572
Quantity 25 g
Molecular Weight (g/mol) 205.3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.