Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(3-Pyridyl)phenylboronic Acid Hydrochloride (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. P23925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23925G 5 g
P23921G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P23925G Supplier TCI America Supplier No. P23925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

Molecular Formula C11H10BNO2
Synonym 4-(3-Pyridyl)benzeneboronic Acid Hydrochloride

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 235.47
Physical Form Crystalline Powder
Chemical Name or Material 4-(3-Pyridyl)phenylboronic Acid Hydrochloride (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.