Learn More
Medchemexpress LLC 4-((3-((R)-3-(3,4-Dimethoxyphenyl)-1-(((S)-1-(3,3-dimethyl-2-oxopentanoyl)piperidine-2-carbonyl)oxy | 1092369-24-8 | MFCD31631580 | 99.6% | 624.72 g·mol⁻¹ | C34H44N2O9 | 50 MG
SDP
Catalog No. 5000241441
Encompass_Preferred
SLF-amido-C2-COOH is a synthetic FKBP12-binding ligand used as a PROTAC building block. It couples an SLF recognition motif to a short linker ending in a carboxylic acid, providing a convenient functional handle for conjugation in bifunctional degrader synthesis.
  • Acts as an FKBP12-binding moiety for PROTAC construction.
  • Provides a carboxylic acid handle for amide or ester coupling.
  • Compatible with standard small-molecule coupling chemistries.
  • High purity suitable for research applications (≈99.6%).
  • Molecular formula C34H44N2O9; molecular weight 624.72 g·mol⁻¹.
  • Supplied as an off-white to white solid for synthetic use.
Catalog No. 50-002-41441 Supplier Medchemexpress LLC Supplier No. HY10745250MG
SLF-amido-C2-COOH is a synthetic FKBP12-binding ligand used as a PROTAC building block. It couples an SLF recognition motif to a short linker ending in a carboxylic acid, providing a convenient functional handle for conjugation in bifunctional degrader synthesis.
  • Acts as an FKBP12-binding moiety for PROTAC construction.
  • Provides a carboxylic acid handle for amide or ester coupling.
  • Compatible with standard small-molecule coupling chemistries.
  • High purity suitable for research applications (≈99.6%).
  • Molecular formula C34H44N2O9; molecular weight 624.72 g·mol⁻¹.
  • Supplied as an off-white to white solid for synthetic use.