Learn More
Medchemexpress LLC 4-(3-tosyl-2-(tosylmethyl)propanoyl)benzoic acid | 124243-00-1 | MFCD28142486 | 500.58 g/mol | C25H24O7S2 | 5 MG
SDP
Catalog No. 5000210455
Encompass_Preferred
4-(3-tosyl-2-(tosylmethyl)propanoyl)benzoic acid is a bis-sulfone acid derivative used as a reactive covalent linker with a free carboxylic acid and two tosyl groups. It is applied in conjugation and linker chemistry, and can undergo Michael-type addition reactions.
  • Bis-sulfone acid with two tosyl groups.
  • Contains a free carboxylic acid for conjugation.
  • Reactive toward Michael-type additions.
  • Used as a bifunctional building block in synthesis and bioconjugation.
Catalog No. 50-002-10455 Supplier Medchemexpress LLC Supplier No. HYW0733825MG
4-(3-tosyl-2-(tosylmethyl)propanoyl)benzoic acid is a bis-sulfone acid derivative used as a reactive covalent linker with a free carboxylic acid and two tosyl groups. It is applied in conjugation and linker chemistry, and can undergo Michael-type addition reactions.
  • Bis-sulfone acid with two tosyl groups.
  • Contains a free carboxylic acid for conjugation.
  • Reactive toward Michael-type additions.
  • Used as a bifunctional building block in synthesis and bioconjugation.