Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(3-tosyl-2-(tosylmethyl)propanoyl)benzoic acid | 124243-00-1 | MFCD28142486 | >97.0% | 500.58 g/mol | C25H24O7S2 | 10 MG
SDP

Catalog No. 5000210289
Encompass_Preferred

4-(3-tosyl-2-(tosylmethyl)propanoyl)benzoic acid is a benzoic acid derivative supplied as a research reagent for biochemical assays and chemical synthesis. It has the molecular formula C25H24O7S2 and a molecular weight of 500.58 g/mol, and is provided as a solid for laboratory use.

  • Benzoic acid derivative used as a biochemical assay reagent.
  • CAS number 124243-00-1.
  • Molecular weight 500.58 g/mol; formula C25H24O7S2.
  • Purity greater than 97.0% as reported by third-party suppliers.
  • Storage: store at room temperature for up to 3 years; in solvent, keep at -80°C for 2 years or -20°C for 1 year.

Catalog No. 50-002-10289 Supplier Medchemexpress LLC Supplier No. HYW07338210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-(3-tosyl-2-(tosylmethyl)propanoyl)benzoic acid is a benzoic acid derivative supplied as a research reagent for biochemical assays and chemical synthesis. It has the molecular formula C25H24O7S2 and a molecular weight of 500.58 g/mol, and is provided as a solid for laboratory use.

  • Benzoic acid derivative used as a biochemical assay reagent.
  • CAS number 124243-00-1.
  • Molecular weight 500.58 g/mol; formula C25H24O7S2.
  • Purity greater than 97.0% as reported by third-party suppliers.
  • Storage: store at room temperature for up to 3 years; in solvent, keep at -80°C for 2 years or -20°C for 1 year.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.