Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-(3-tosyl-2-(tosylmethyl)propanoyl)benzoic acid | 124243-00-1 | MFCD28142486 | >97.0% | 500.58 g/mol | C25H24O7S2 | 50 MG
SDP

Catalog No. 5000210290
Encompass_Preferred

4-(3-tosyl-2-(tosylmethyl)propanoyl)benzoic acid is a tosylated benzoic-acid derivative used as a research chemical and biochemical assay reagent. It contains a free carboxylic acid and two tosyl groups and is typically supplied in small milligram quantities for synthetic and assay applications.

  • Contains two tosyl groups and a free carboxylic acid.
  • Used as a biochemical assay reagent and synthetic intermediate.
  • Typical purity around 97% (batch-dependent).
  • Molecular weight 500.58 g/mol; chemical formula C25H24O7S2.
  • Available in small milligram pack sizes for research use.

Catalog No. 50-002-10290 Supplier Medchemexpress LLC Supplier No. HYW07338250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-(3-tosyl-2-(tosylmethyl)propanoyl)benzoic acid is a tosylated benzoic-acid derivative used as a research chemical and biochemical assay reagent. It contains a free carboxylic acid and two tosyl groups and is typically supplied in small milligram quantities for synthetic and assay applications.

  • Contains two tosyl groups and a free carboxylic acid.
  • Used as a biochemical assay reagent and synthetic intermediate.
  • Typical purity around 97% (batch-dependent).
  • Molecular weight 500.58 g/mol; chemical formula C25H24O7S2.
  • Available in small milligram pack sizes for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.