Learn More
Medchemexpress LLC 4-(3-tosyl-2-(tosylmethyl)propanoyl)benzoic acid (bis-sulfone acid) | 124243-00-1 | MFCD28142486 | 97.7% | 500.58 g/mol | C25H24O7S2 | 25 MG
SDP
Catalog No. 5000208869
Encompass_Preferred
4-(3-tosyl-2-(tosylmethyl)propanoyl)benzoic acid is a bifunctional bis-sulfone carboxylic acid used as a covalent linker in conjugation chemistry. It contains a free carboxylic acid and two tosyl groups that can participate in Michael-type additions, enabling site-selective conjugation and linker installation for small-molecule and bioconjugation applications.
  • contains a free carboxylic acid for conjugation
  • two tosyl groups enable Michael-type additions
  • high purity suitable for research applications
  • available in multiple small sizes for laboratory use
Catalog No. 50-002-08869 Supplier Medchemexpress LLC Supplier No. 25MGHYW073382
4-(3-tosyl-2-(tosylmethyl)propanoyl)benzoic acid is a bifunctional bis-sulfone carboxylic acid used as a covalent linker in conjugation chemistry. It contains a free carboxylic acid and two tosyl groups that can participate in Michael-type additions, enabling site-selective conjugation and linker installation for small-molecule and bioconjugation applications.
  • contains a free carboxylic acid for conjugation
  • two tosyl groups enable Michael-type additions
  • high purity suitable for research applications
  • available in multiple small sizes for laboratory use