Learn More
Medchemexpress LLC 4-[(3aS,4R,9bR)-6-ethoxy-3a,4,5,9b-tetrahydro-3H-cyclopenta[c]quinolin-4-yl]benzoic acid | 1357248-83-9 | 98.7% | 335.40 g/mol | C21H21NO3 | 10MM 1ML
SDP
Supplier:  Medchemexpress LLC HY12390510MM 1ML
Encompass_Preferred
LIN28 inhibitor LI71 is a research small-molecule inhibitor that disrupts LIN28:let-7 binding and LIN28-mediated oligouridylation. It is used in LIN28-driven cancer research and related biochemical studies, and has demonstrated activity against Plasmodium falciparum cold shock protein interactions. Available as a 10 mM solution in DMSO and as multiple solid pack sizes.
Catalog No. 50-002-35387