Learn More
Medchemexpress LLC 4(3H)-pteridinone, 2-amino-6-methyl- | 708-75-8 | MFCD00012137 | 99.8% | 177.16 g/mol | C7H7N5O | 250 MG
SDP
Catalog No. 5000303429
Encompass_Preferred
6-Methylpterin is a pterin derivative used in biochemical research related to folate metabolism and photochemical studies. It produces singlet oxygen and hydrogen peroxide upon light exposure and is employed as a probe and analytical standard. Supplied as a white to yellow solid with high purity, the compound requires light-protected storage and defined solvent storage conditions to maintain stability.
  • High purity (HPLC) 99.84% for reliable experimental results.
  • Well-characterized by 1H NMR and mass spectrometry, confirming identity.
  • Stable solid form; store protected from light at 4°C.
  • Solvent storage guidelines: -80°C (6 months) or -20°C (1 month) to preserve stability.
  • Suitable for folate-related biochemical assays and photochemical studies.
Catalog No. 50-003-03429 Supplier Medchemexpress LLC Supplier No. 250MGHYW037819
6-Methylpterin is a pterin derivative used in biochemical research related to folate metabolism and photochemical studies. It produces singlet oxygen and hydrogen peroxide upon light exposure and is employed as a probe and analytical standard. Supplied as a white to yellow solid with high purity, the compound requires light-protected storage and defined solvent storage conditions to maintain stability.
  • High purity (HPLC) 99.84% for reliable experimental results.
  • Well-characterized by 1H NMR and mass spectrometry, confirming identity.
  • Stable solid form; store protected from light at 4°C.
  • Solvent storage guidelines: -80°C (6 months) or -20°C (1 month) to preserve stability.
  • Suitable for folate-related biochemical assays and photochemical studies.